• USA Home
  • 214108 - N,N′-Bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea dipropanoate

EMAIL THIS PAGE TO A FRIEND
214108 Sigma-Aldrich

N,N′-Bis[3-(4,5-dihydro-1H-imidazol-2-yl)phenyl]urea dipropanoate

99%

Purchase

Properties

assay   99%
mp   205 °C (dec.) (lit.)
SMILES string   CCC(O)=O.CCC(O)=O.O=C(Nc1cccc(c1)C2=NCCN2)Nc3cccc(c3)C4=NCCN4
InChI   1S/C19H20N6O.2C3H6O2/c26-19(24-15-5-1-3-13(11-15)17-20-7-8-21-17)25-16-6-2-4-14(12-16)18-22-9-10-23-18;2*1-2-3(4)5/h1-6,11-12H,7-10H2,(H,20,21)(H,22,23)(H2,24,25,26);2*2H2,1H3,(H,4,5)
InChI key   AFGQXWSHYUHHNV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

33441 Imidocarb dipropionate, VETRANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?