EMAIL THIS PAGE TO A FRIEND
215732 Sigma-Aldrich

Disperse Orange 25

Dye content 20 %

Synonym: 3-[N-Ethyl-4-(4-nitrophenylazo)phenylamino]propionitrile

Purchase

Properties

composition   Dye content, 20%
mp   170 °C (dec.) (lit.)
λmax   455 nm
SMILES string   CCN(CCC#N)c1ccc(cc1)\N=N\c2ccc(cc2)[N+]([O-])=O
InChI   1S/C17H17N5O2/c1-2-21(13-3-12-18)16-8-4-14(5-9-16)19-20-15-6-10-17(11-7-15)22(23)24/h4-11H,2-3,13H2,1H3/b20-19+
InChI key   ZSPPPAFDNHYXNW-FMQUCBEESA-N

Description

Features and Benefits

Adsorptive incorporation into molecular sieves yielded nanocomposites with nonlinear and photosensitive optical properties.

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
TZ4700000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

364819 Disperse Orange 25, Dye content 95 %

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?