• USA Home
  • 217026 - 4-Amino-4′-methoxydiphenylamine hydrochloride

EMAIL THIS PAGE TO A FRIEND
217026 Sigma-Aldrich

4-Amino-4′-methoxydiphenylamine hydrochloride

97%

Synonym: N-(p-Methoxyphenyl)-p-phenylenediamine, Variamine Blue B Base

Purchase

Properties

assay   97%
mp   249-255 °C (lit.)
SMILES string   Cl.COc1ccc(Nc2ccc(N)cc2)cc1
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?