• USA Home
  • 217980 - Vincristine sulfate salt

EMAIL THIS PAGE TO A FRIEND
217980 Sigma-Aldrich

Vincristine sulfate salt

Synonym: 22-Oxovincaleukoblastine sulfate salt, Leurocristine sulfate salt, VCR

  • CAS Number 2068-78-2

  • Empirical Formula (Hill Notation) C46H56N4O10 · H2SO4

  • Molecular Weight 923.04

  •  Beilstein/REAXYS Number 3924631

Purchase
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?