• USA Home
  • 219053 - 2′,5′-Dimethoxy-4-stilbenamine

EMAIL THIS PAGE TO A FRIEND
219053

2′,5′-Dimethoxy-4-stilbenamine

97%

Synonym: DMBAS

  • CAS Number 32180-65-7

  • Empirical Formula (Hill Notation) C16H17NO2

  • Molecular Weight 255.31

  •  MDL number MFCD00007915

Purchase

Properties

assay   97%
SMILES string   COc1ccc(OC)c(\C=C\c2ccc(N)cc2)c1
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?