• USA Home
  • 219436 - m-Fluoro-DL-phenylalanine

EMAIL THIS PAGE TO A FRIEND
219436 Aldrich

m-Fluoro-DL-phenylalanine

95%

  • CAS Number 2629-54-1

  • Linear Formula FC6H4CH2CH(NH2)COOH

  • Molecular Weight 183.18

  •  Beilstein/REAXYS Number 2939807

  •  MDL number MFCD00004273

Purchase

Properties

assay   95%
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?