• USA Home
  • 219711 - D-Histidine monohydrochloride monohydrate

EMAIL THIS PAGE TO A FRIEND
219711 Sigma-Aldrich

D-Histidine monohydrochloride monohydrate

99%

Synonym: D-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride

  • CAS Number 6341-24-8

  • Empirical Formula (Hill Notation) C6H9N3O2 · HCl · H2O

  • Molecular Weight 209.63

  •  Beilstein/REAXYS Number 4161356

  •  EC Number 228-733-3

  •  MDL number MFCD00150081

Purchase

Properties

InChI Key   CMXXUDSWGMGYLZ-ZJIMSODOSA-N
assay   99%
optical activity   [α]25/D −10.0°, c = 2 in 5 M HCl
mp   254 °C (dec.) (lit.)

Description

Other Notes

Unnatural isomer of histidine.

Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H7625 D-Histidine monohydrochloride monohydrate, ≥98% (TLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?