• USA Home
  • 219959 - cis-4-Hydroxy-L-proline

EMAIL THIS PAGE TO A FRIEND
219959 Aldrich

cis-4-Hydroxy-L-proline

99%

Synonym: (2S,4S)-(−)-4-Hydroxy-2-pyrrolidinecarboxylic acid, CHP

  • CAS Number 618-27-9

  • Empirical Formula (Hill Notation) C5H9NO3

  • Molecular Weight 131.13

  •  Beilstein/REAXYS Number 81440

  •  EC Number 210-542-1

  •  MDL number MFCD00064319

Purchase

Properties

InChI Key   PMMYEEVYMWASQN-IMJSIDKUSA-N
assay   99%
optical activity   [α]20/D −59.0°, c = 2 in H2O
mp   257 °C (dec.) (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H1637 cis-4-Hydroxy-L-proline, collagen synthesis inhibitor

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?