• USA Home
  • 220302 - (Iodomethyl)trimethylsilane

EMAIL THIS PAGE TO A FRIEND
220302 Sigma-Aldrich

(Iodomethyl)trimethylsilane

99%

Synonym: (Trimethylsilyl)methyl iodide

Purchase

Properties

assay   99%
contains   copper as stabilizer
refractive index   n20/D 1.491 (lit.)
bp   139-141 °C (lit.)
density   1.443 g/mL at 25 °C (lit.)
SMILES string   C[Si](C)(C)CI
InChI   1S/C4H11ISi/c1-6(2,3)4-5/h4H2,1-3H3
InChI key   VZNYXGQMDSRJAL-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2924 8(3) / PGIII
WGK Germany 
3
Flash Point(F) 
84.2 °F
Flash Point(C) 
29 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

57903 (Iodomethyl)trimethylsilane, ≥99.0% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?