• USA Home
  • 221112 - N-(4-Nitrobenzoyl)-L-glutamic acid hemihydrate

EMAIL THIS PAGE TO A FRIEND
221112 Sigma-Aldrich

N-(4-Nitrobenzoyl)-L-glutamic acid hemihydrate

98%

  • CAS Number 6758-40-3

  • Linear Formula HO2CCH2CH2CH(NHCOC6H4NO2)CO2H · 1/2H2O

  • Molecular Weight 305.24

  •  EC Number 229-820-9

  •  PubChem Substance ID 24853214

Purchase

Properties

assay   98%
optical activity   [α]24/D +15.1°, c = 2 in 1 M NaOH
mp   117-118 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?