• USA Home
  • 221554 - 1-Heptanesulfonic acid sodium salt

EMAIL THIS PAGE TO A FRIEND
221554 Sigma-Aldrich

1-Heptanesulfonic acid sodium salt

98%

Synonym: Sodium 1-heptanesulfonate

Purchase

Properties

assay   98%
mp   >300 °C (lit.)
SMILES string   [Na+].CCCCCCCS([O-])(=O)=O

Description

Other Notes

This product has been replaced by H2766-Sigma-Aldrich | Sodium 1-heptanesulfonate

Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

H2766 Sodium 1-heptanesulfonate

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?