• USA Home
  • 222208 - 5,6,8,9-Tetrahydrobenz[a]anthracen-11(10H)-one

EMAIL THIS PAGE TO A FRIEND
222208 Sigma-Aldrich

5,6,8,9-Tetrahydrobenz[a]anthracen-11(10H)-one

99%

  • CAS Number 1470-04-8

  • Empirical Formula (Hill Notation) C18H16O

  • Molecular Weight 248.32

  •  MDL number MFCD00003597

Purchase

Properties

assay   99%
SMILES string   O=C1CCCc2cc3CCc4ccccc4-c3cc12
InChI   1S/C18H16O/c19-18-7-3-5-13-10-14-9-8-12-4-1-2-6-15(12)16(14)11-17(13)18/h1-2,4,6,10-11H,3,5,7-9H2
InChI key   YVGXWBAAHRAVFX-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?