• USA Home
  • 222631 - syn-4-Fluorobenzaldoxime

EMAIL THIS PAGE TO A FRIEND
222631 Sigma-Aldrich

syn-4-Fluorobenzaldoxime

97%

Purchase

Properties

assay   97%
mp   82-85 °C (lit.)
SMILES string   O\N=C\c1ccc(F)cc1
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
CU6260000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?