• USA Home
  • 227536 - 4-Methyl-2,6-dinitrophenol

EMAIL THIS PAGE TO A FRIEND
227536 Sigma-Aldrich

4-Methyl-2,6-dinitrophenol

90%

Synonym: 2,6-Dinitro-4-methylphenol, 2,6-Dinitro-p-cresol, 3,5-Dinitro-4-hydroxytoluene

  • CAS Number 609-93-8

  • Linear Formula CH3C6H2(NO2)2OH

  • Molecular Weight 198.13

  •  Beilstein/REAXYS Number 1978786

  •  EC Number 210-203-8

  •  MDL number MFCD00007121

  •  PubChem Substance ID 24853638

Purchase

Properties

assay   90%
contains   10% water
mp   76-80 °C (lit.)
SMILES string   Cc1cc(c(O)c(c1)[N+]([O-])=O)[N+]([O-])=O
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
GO9800000
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?