• USA Home
  • 227943 - 1,3,5-Triisopropylcyclohexane,mixture of isomers

EMAIL THIS PAGE TO A FRIEND
227943

1,3,5-Triisopropylcyclohexane,mixture of isomers

98%

  • CAS Number 34387-60-5

  • Empirical Formula (Hill Notation) C15H30

  • Molecular Weight 210.40

  •  MDL number MFCD00001485

Purchase

Properties

assay   98%
SMILES string   CC(C)C1CC(CC(C1)C(C)C)C(C)C

Description

Application

Useful for calorimetric heat of immersion studies.

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?