• USA Home
  • 228567 - 5-Chloro-2-methoxyaniline

EMAIL THIS PAGE TO A FRIEND
228567 Aldrich

5-Chloro-2-methoxyaniline

98%

Synonym: 2-Amino-4-chloroanisole, 5-Chloro-o-anisidine

  • CAS Number 95-03-4

  • Linear Formula ClC6H3(OCH3)NH2

  • Molecular Weight 157.60

  •  Beilstein/REAXYS Number 879161

  •  EC Number 202-385-2

  •  MDL number MFCD00007777

Purchase

Properties

assay   98%
mp   81-83 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 2233 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
276.8 °F
Flash Point(C) 
136 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?