EMAIL THIS PAGE TO A FRIEND
228869 Sigma-Aldrich

Indocyanine green

Dye content 80 %

Synonym: Cardiogreen, Foxgreen, IR-125

  • CAS Number 3599-32-4

  • Empirical Formula (Hill Notation) C43H47N2NaO6S2

  • Molecular Weight 774.96

  •  Beilstein/REAXYS Number 4115884

  •  EC Number 222-751-5

  •  MDL number MFCD00013078

Purchase

Properties

composition   Dye content, 80%
λmax   775 nm
SMILES string   [Na+].CC1(C)/C(=C\C=C\C=C\C=C\C2=[N+](CCCCS([O-])(=O)=O)c3ccc4ccccc4c3C2(C)C)N(CCCCS([O-])(=O)=O)c5ccc6ccccc6c15

Description

Application

IR laser dye.

Other Notes

This product has been replaced by I2633-SIGMA | Cardiogreen polymethine dye

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

I2633 Cardiogreen, polymethine dye

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?