EMAIL THIS PAGE TO A FRIEND
228923 Sigma-Aldrich

Ammonium L-tartrate

99%

Synonym: L-Tartaric acid ammonium salt, Ammonium tartrate dibasic, Diammonium tartrate, L(+)-Tartaric acid diammonium salt

  • CAS Number 3164-29-2

  • Linear Formula (NH4)2C4H4O6

  • Molecular Weight 184.15

  •  Beilstein/REAXYS Number 6120352

  •  EC Number 221-618-9

  •  MDL number MFCD00013073

Purchase

Properties

assay   99%
optical activity   [α]15/D +32.4°, c = 1.84 in H2O
density   1.601 g/mL at 25 °C (lit.)
SMILES string   N.N.O[C@H]([C@@H](O)C(O)=O)C(O)=O
Safety & Documentation

Safety Information

Personal Protective Equipment 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?