• USA Home
  • 22995 - trans-3-Chloroacrylic acid

EMAIL THIS PAGE TO A FRIEND
22995 Aldrich

trans-3-Chloroacrylic acid

≥98.0% (HPLC)

Purchase

Properties

InChI Key   MHMUCYJKZUZMNJ-OWOJBTEDSA-N
assay   ≥98.0% (HPLC)
mp   84-86 °C
storage temp.   2-8°C
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
RTECS 
AS5960000

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?