• USA Home
  • 230332 - (−)-Isopropyl L-lactate

EMAIL THIS PAGE TO A FRIEND
230332 Sigma-Aldrich

(−)-Isopropyl L-lactate

99%

  • CAS Number 63697-00-7

  • Linear Formula CH3CH(OH)CO2CH(CH3)2

  • Molecular Weight 132.16

  •  Beilstein/REAXYS Number 3601100

  •  EC Number 264-417-1

  •  MDL number MFCD00009728

Purchase

Properties

vapor density   4.2 (vs air)
assay   99%
optical activity   [α]25/D −10.3°, neat
refractive index   n20/D 1.411 (lit.)
bp   166-168 °C (lit.)
density   0.988 g/mL at 25 °C (lit.)
SMILES string   CC(C)OC(=O)[C@H](C)O
Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
134.6 °F
Flash Point(C) 
57 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?