• USA Home
  • 23043 - Dichloro(2-chloroethyl)methylsilane

EMAIL THIS PAGE TO A FRIEND
23043 Aldrich

Dichloro(2-chloroethyl)methylsilane

≥95.0% (GC)

Synonym: (2-Chloroethyl)-dichloro-methylsilane, (2-Chloroethyl)methyl-dichlorosilane

  • CAS Number 7787-85-1

  • Empirical Formula (Hill Notation) C3H7Cl3Si

  • Molecular Weight 177.53

  •  Beilstein Registry Number 1734992

  •  EC Number 232-134-2

  •  MDL number MFCD00053237

Purchase

Properties

assay   ≥95.0% (GC)
refractive index   n20/D 1.458(lit.)
density   1.241 g/mL at 20 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2986 3(8) / PGII
WGK Germany 
3
Flash Point(F) 
89.6 °F
Flash Point(C) 
32 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?