EMAIL THIS PAGE TO A FRIEND
232300 Aldrich

1,2-Benzisoxazole

95%

  • CAS Number 271-95-4

  • Empirical Formula (Hill Notation) C7H5NO

  • Molecular Weight 119.12

  •  Beilstein/REAXYS Number 2154

  •  EC Number 205-983-1

  •  MDL number MFCD00005852

Purchase

Properties

InChI Key   KTZQTRPPVKQPFO-UHFFFAOYSA-N
assay   95%
refractive index   n20/D 1.561 (lit.)
bp   90-92 °C/15 mmHg(lit.)
density   1.174 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
186.8 °F
Flash Point(C) 
86 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

12255 1,2-Benzisoxazole, ≥95.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?