• USA Home
  • 232351 - S-(tert-Butylthio)-L-cysteine hydrate

EMAIL THIS PAGE TO A FRIEND
232351 Sigma-Aldrich

S-(tert-Butylthio)-L-cysteine hydrate

98%

Synonym: 3-(tert-Butyldithio)-L-alanine

Purchase

Properties

assay   98%
optical activity   [α]21/D −82°, c = 2 in 1 M HCl
mp   177 °C (dec.) (lit.)
SMILES string   CC(C)(C)SSC[C@H](N)C(O)=O
InChI   1S/C7H15NO2S2/c1-7(2,3)12-11-4-5(8)6(9)10/h5H,4,8H2,1-3H3,(H,9,10)/t5-/m0/s1
InChI key   TWMBHZTWEDJDRC-YFKPBYRVSA-N
Safety & Documentation

Safety Information

WGK Germany 
3
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B5890 S-tert-Butylmercapto-L-cysteine

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?