• USA Home
  • 232572 - S,S′-Ethylene di(p-toluenethiosulfonate)

EMAIL THIS PAGE TO A FRIEND
232572 Sigma-Aldrich

S,S′-Ethylene di(p-toluenethiosulfonate)

99%

Synonym: 1,2-Di(p-tosylthio)ethane, 1,2-Ethanedithiol ditosylate, Ethylene di(thiotosylate)

  • CAS Number 2225-23-2

  • Linear Formula CH3C6H4SO2SCH2CH2SSO2C6H4CH3

  • Molecular Weight 402.57

  •  Beilstein/REAXYS Number 2018502

  •  EC Number 218-752-5

  •  MDL number MFCD00008550

Purchase

Properties

assay   99%
mp   72-75 °C (lit.)
SMILES string   Cc1ccc(cc1)S(=O)(=O)SCCSS(=O)(=O)c2ccc(C)cc2
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?