• USA Home
  • 232718 - Dibutylboryl trifluoromethanesulfonate solution

EMAIL THIS PAGE TO A FRIEND
232718 Sigma-Aldrich

Dibutylboryl trifluoromethanesulfonate solution

1.0 M in diethyl ether

Synonym: Dibutylboron triflate solution

  • CAS Number 60669-69-4

  • Linear Formula CF3SO3B[(CH2)3CH3]2

  • Molecular Weight 274.11

  •  Beilstein/REAXYS Number 1968660

  •  MDL number MFCD00009669

  •  PubChem Substance ID 24853816

Purchase

Properties

concentration   1.0 M in diethyl ether
density   0.815 g/mL at 25 °C
SMILES string   CCCCB(CCCC)OS(=O)(=O)C(F)(F)F
InChI   1S/C9H18BF3O3S/c1-3-5-7-10(8-6-4-2)16-17(14,15)9(11,12)13/h3-8H2,1-2H3
InChI key   FAVAVMFXAKZTMV-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
May form explosive peroxides.
RIDADR 
UN 2924 8(3) / PGI
WGK Germany 
3
Flash Point(F) 
-40 °F
Flash Point(C) 
-40 °C

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?