• USA Home
  • 233218 - 3,5-Bis(trifluoromethyl)benzonitrile

EMAIL THIS PAGE TO A FRIEND
233218 Sigma-Aldrich

3,5-Bis(trifluoromethyl)benzonitrile

97%

Purchase

Properties

assay   97%
refractive index   n20/D 1.4175 (lit.)
density   1.42 g/mL at 25 °C (lit.)
SMILES string   FC(F)(F)c1cc(cc(c1)C(F)(F)F)C#N
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
163.4 °F
Flash Point(C) 
73 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?