• USA Home
  • 233463 - 5-Methyl-2-thiouracil

EMAIL THIS PAGE TO A FRIEND
233463 Sigma-Aldrich

5-Methyl-2-thiouracil

98%

Synonym: 4-Hydroxy-2-mercapto-5-methylpyrimidine

  • CAS Number 636-26-0

  • Empirical Formula (Hill Notation) C5H6N2OS

  • Molecular Weight 142.18

  •  MDL number MFCD00009777

  •  PubChem Substance ID 329752412

Purchase

Properties

InChI Key   ZLAQATDNGLKIEV-UHFFFAOYSA-N
assay   98%
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
XP2108500

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?