• USA Home
  • 233986 - PIPES disodium salt hydrate

EMAIL THIS PAGE TO A FRIEND
233986 Sigma-Aldrich

PIPES disodium salt hydrate

98%

  • CAS Number 331717-47-6

  • Empirical Formula (Hill Notation) C8H16N2Na2O6S2 · xH2O

  • Molecular Weight 346.33 (anhydrous basis)

  •  EC Number 278-562-3

  •  MDL number MFCD00166972

  •  PubChem Substance ID 24854015

Purchase

Properties

assay   98%
SMILES string   [Na+].[Na+].[H]O[H].[O-]S(=O)(=O)CCN1CCN(CC1)CCS([O-])(=O)=O
InChI   1S/C8H18N2O6S2.2Na.H2O/c11-17(12,13)7-5-9-1-2-10(4-3-9)6-8-18(14,15)16;;;/h1-8H2,(H,11,12,13)(H,14,15,16);;;1H2/q;2*+1;/p-2
InChI key   JOHAUORJTCNVKB-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Buffers - From Bnech to Bulk
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

P3768 PIPES disodium salt, ≥99% (titration)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?