• USA Home
  • 235415 - Ethyl 1,3-dithiane-2-carboxylate

EMAIL THIS PAGE TO A FRIEND
235415 Sigma-Aldrich

Ethyl 1,3-dithiane-2-carboxylate

97%

Synonym: Glyoxylic acid ethyl ester trimethylenemercaptal

  • CAS Number 20462-00-4

  • Empirical Formula (Hill Notation) C7H12O2S2

  • Molecular Weight 192.30

  •  Beilstein/REAXYS Number 1424352

  •  EC Number 243-838-4

  •  MDL number MFCD00006657

Purchase

Properties

assay   97%
refractive index   n20/D 1.539 (lit.)
bp   75-77 °C/0.2 mmHg (lit.)
density   1.22 g/mL at 25 °C (lit.)
SMILES string   CCOC(=O)C1SCCCS1
InChI   1S/C7H12O2S2/c1-2-9-6(8)7-10-4-3-5-11-7/h7H,2-5H2,1H3
InChI key   ANEDZEVDORCLPM-UHFFFAOYSA-N

Description

Application

Reagent for the synthesis of α-ketoesters.

Safety & Documentation

Safety Information

Symbol 
GHS02  GHS02
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3272 3 / PGIII
WGK Germany 
3
Flash Point(F) 
131 °F
Flash Point(C) 
55 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

43749 Ethyl 1,3-dithiane-2-carboxylate, technical, ≥90% (GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?