• USA Home
  • 235458 - Methyl pentadecanoate

EMAIL THIS PAGE TO A FRIEND
235458 Sigma-Aldrich

Methyl pentadecanoate

98%

Synonym: Pentadecanoic acid methyl ester

Purchase

Properties

InChI Key   XIUXKAZJZFLLDQ-UHFFFAOYSA-N
assay   98%
refractive index   n20/D 1.438 (lit.)
bp   141-142 °C/3 mmHg(lit.)
mp   18-19 °C (lit.)
density   0.862 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
>235.4 °F
Flash Point(C) 
>113 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

P6250 Methyl pentadecanoate, ≥98.5%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?