• USA Home
  • 235792 - 3-(Trimethoxysilyl)propyl methacrylate

EMAIL THIS PAGE TO A FRIEND
235792 Aldrich

3-(Trimethoxysilyl)propyl methacrylate

97%

Synonym: Silane A174, [3-(Methacryloyloxy)propyl]trimethoxysilane

  • CAS Number 2530-85-0

  • Linear Formula H2C=C(CH3)CO2(CH2)3Si(OCH3)3

  • Molecular Weight 248.35

  •  Beilstein Registry Number 1952435

  •  EC Number 219-785-8

  •  MDL number MFCD00008593

  •  PubChem Substance ID 329752431

Purchase

Properties

InChI Key   XDLMVUHYZWKMMD-UHFFFAOYSA-N
assay   97%
form   liquid
impurities   1% methyl alcohol from hydrolysis
refractive index   n20/D 1.431(lit.)
bp   190 °C(lit.)
density   1.045 g/mL at 25 °C(lit.)

Description

Application

Useful as a monomer in the formation of sol-gel composites.

Safety & Documentation
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?