• USA Home
  • 236934 - Dimethyl sulfoxide-d6

EMAIL THIS PAGE TO A FRIEND
236934 Sigma-Aldrich

Dimethyl sulfoxide-d6

"100%", 99.96 atom % D

Synonym: (Methyl sulfoxide)-d6, DMSO-d6, Hexadeuterodimethyl sulfoxide

Purchase

Properties

Related Categories Dimethyl sulfoxide-d6, NMR Solvents, Solvent by Application, Solvents, Spectroscopy Solvents (IR, NMR, UV/Vis) More...
vapor pressure   0.42 mmHg ( 20 °C)
InChI Key   IAZDPXIOMUYVGZ-WFGJKAKNSA-N
isotopic purity   99.96 atom % D
assay   ≥99.0%
autoignition temp.   573 °F
expl. lim.   42 %
packaging   pk of 10 × 0.5 mL
impurities   ≤0.0250% water
water
refractive index   n20/D 1.476(lit.)
bp   189 °C(lit.)
mp   20.2 °C (lit.)
density   1.190 g/mL at 25 °C(lit.)
mass shift   M+6
Safety & Documentation

Safety Information

WGK Germany 
2
Flash Point(F) 
188.6 °F
Flash Point(C) 
87 °C
Protocols & Articles

Articles

Pressure-Temperature Calculator for Solvents

*Calculations provide estimated results based on Antoine's Coefficients and their associated temperature range values. Antoine's Coefficients were obtained from the National Institute of Standards an...

Related Content

Pressure-Temperature Nomograph Interactive Tool

The Pressure-Temperature Nomograph tool is an application of the Clausius-Clapeyron Equation, which assumes the heat of vaporization is a constant over a pressure range. Antoine´s Equation gets aroun...
Keywords: Evaporation, Vaporization

Properties of Solvents Table

a. Gant, R., LC/GC 10 (7), 515 (1992) 1. At 20 °C 2. At 20 °C +/- 5 °C relative to water at 4 °C 3. At 10 mmHg pressure 4. At l=589.32 nm
Keywords: Gas chromatography, High performance liquid chromatography

Peer-Reviewed Papers
15

References

Related Products

accessory

Product #

Description

Add to Cart

Z693847 Scienceware® Break-Safe ampule opener

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?