• USA Home
  • 237663 - (+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane

EMAIL THIS PAGE TO A FRIEND
237663 Sigma-Aldrich

(+)-2,3-O-Isopropylidene-2,3-dihydroxy-1,4-bis(diphenylphosphino)butane

98%

Synonym: (+)-DIOP, (+)-1,4-Bis(diphenylphosphino)-1,4-dideoxy-2,3-O-isopropylidene-D-threitol, (4S,5S)-4,5-Bis(diphenylphosphinomethyl)-2,2-dimethyl-1,3-dioxolane, (4S-trans)-[(2,2-Dimethyl-1,3-dioxolane-4,5-diyl)bis(methylene)]bis(diphenylphosphine), (S,S)-DIOP

  • CAS Number 37002-48-5

  • Empirical Formula (Hill Notation) C31H32O2P2

  • Molecular Weight 498.53

  •  Beilstein/REAXYS Number 1630280

  •  EC Number 253-307-9

  •  MDL number MFCD00009760

  •  PubChem Substance ID 24854262

Purchase

Properties

Related Categories Asymmetric Synthesis, Chemical Synthesis, Chiral Catalysts, Ligands, and Reagents, DIOP, Privileged Ligands and Complexes More...
assay   98%
optical activity   [α]19/D +26°, c = 2.3 in chloroform
mp   86-88 °C (lit.)
storage temp.   2-8°C
SMILES string   CC1(C)O[C@H](CP(c2ccccc2)c3ccccc3)[C@@H](CP(c4ccccc4)c5ccccc5)O1
InChI   1S/C31H32O2P2/c1-31(2)32-29(23-34(25-15-7-3-8-16-25)26-17-9-4-10-18-26)30(33-31)24-35(27-19-11-5-12-20-27)28-21-13-6-14-22-28/h3-22,29-30H,23-24H2,1-2H3/t29-,30-/m1/s1
InChI key   VCHDBLPQYJAQSQ-LOYHVIPDSA-N

Description

Application

For the in situ preparation of chiral hydrogenation catalysts.

Ligand for Rh(I)-catalyzed asymmetric hydrogenation of enamides, 1,4-disilylation of α,β-unsaturated ketones, Heck reaction of alkenyl iodides, and hydroformylation of olefins.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Articles

DIOP

DIOP is one of the first chiral ligands used for asymmetric hydrogenation catalysis. DIOP can be synthesized in four simple steps from tartaric acid. Chelation with a rhodium complex generates an opt...
William Sommer and Daniel Weibel
ChemFiles 2008, 8.2, 81.
Keywords: Asymmetric synthesis, Catalysis, Hydrogenations, Ligands

DIPAMP

Rewarded by a Nobel Prize in 2001 for his pioneering work in asymmetric synthesis, Knowles was the first to develop a transition-metal chiral catalyst based on a chiral diphosphine ligand, DIPAMP, th...
William Sommer and Daniel Weibel
ChemFiles 2008, 8.2, 81.
Keywords: Asymmetric synthesis, Catalysis, Hydrogenations, Ligands, Parkinson Disease

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?