• USA Home
  • 237671 - DL-2-Amino-1-hexanol

EMAIL THIS PAGE TO A FRIEND
237671 Sigma-Aldrich

DL-2-Amino-1-hexanol

90%, technical grade

Synonym: DL-Norleucinol

Purchase

Properties

Related Categories Amino Alcohols, Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds More...
grade   technical grade
InChI Key   DPEOTCPCYHSVTC-UHFFFAOYSA-N
assay   90%
refractive index   n20/D 1.4522 (lit.)
bp   190-192 °C/740 mmHg(lit.)
density   0.912 g/mL at 25 °C (lit.)

Description

General description

Reduced mobility of protonated DL-2-amino-1-hexanol in air, at 200°C, has been evaluated by ion mobility spectrometry (IMS).

Packaging

1 g in glass bottle

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
212 °F
Flash Point(C) 
100 °C

Documents

Certificate of Analysis

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?