• USA Home
  • 237841 - Tetraheptylammonium bromide

EMAIL THIS PAGE TO A FRIEND
237841 Aldrich

Tetraheptylammonium bromide

98%

  • CAS Number 4368-51-8

  • Linear Formula [CH3(CH2)6]4N(Br)

  • Molecular Weight 490.69

  •  Beilstein Registry Number 3763391

  •  EC Number 224-459-3

  •  MDL number MFCD00011861

Purchase

Properties

InChI Key   YQIVQBMEBZGFBY-UHFFFAOYSA-M
assay   98%
mp   87-89 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?