EMAIL THIS PAGE TO A FRIEND
238341 Sigma-Aldrich

Bromopyruvic acid

98%

Synonym: 3-Bromo-2-oxopropionic acid

Purchase

Properties

assay   98%
SMILES string   OC(=O)C(=O)CBr
InChI   1S/C3H3BrO3/c4-1-2(5)3(6)7/h1H2,(H,6,7)
InChI key   PRRZDZJYSJLDBS-UHFFFAOYSA-N

Description

Application

Substrate employed in the synthesis of substituted pyranones.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3261 8 / PGII
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

16490 Bromopyruvic acid, ≥98.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?