EMAIL THIS PAGE TO A FRIEND
239445 Sigma-Aldrich

Ethyl phenylacetate

≥99%

  • CAS Number 101-97-3

  • Linear Formula C6H5CH2COOC2H5

  • Molecular Weight 164.20

  •  Beilstein/REAXYS Number 509140

  •  EC Number 202-993-8

  •  MDL number MFCD00009178

  •  PubChem Substance ID 24854377

Purchase

Properties

InChI Key   DULCUDSUACXJJC-UHFFFAOYSA-N
assay   ≥99%
refractive index   n20/D 1.497 (lit.)
bp   229 °C(lit.)
density   1.03 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
AJ2824000
Flash Point(F) 
208.4 °F
Flash Point(C) 
98 °C

Documents

Certificate of Analysis

???PDP_SDS_LOT_NUMBER_ERROR_MSG???
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?