EMAIL THIS PAGE TO A FRIEND
239712 Sigma-Aldrich

Tripropylamine

≥99%

  • CAS Number 102-69-2

  • Linear Formula (CH3CH2CH2)3N

  • Molecular Weight 143.27

  •  Beilstein/REAXYS Number 1098331

  •  EC Number 203-047-7

  •  MDL number MFCD00009363

  •  PubChem Substance ID 24854388

Purchase

Properties

vapor density   4.9 (vs air)
vapor pressure   2.9 mmHg ( 20 °C)
InChI Key   YFTHZRPMJXBUME-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   356 °F
expl. lim.   5.6 %
refractive index   n20/D 1.416 (lit.)
bp   155-158 °C(lit.)
  40-42 °C/11 mmHg(lit.)
mp   −93.5 °C (lit.)
density   0.753 g/mL at 25 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
RIDADR 
UN 2260 8(3) / PGIII
WGK Germany 
2
RTECS 
TX1575000
Flash Point(F) 
93.2 °F
Flash Point(C) 
34 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?