EMAIL THIS PAGE TO A FRIEND
240001

Dibutylamine

≥99%

  • CAS Number 111-92-2

  • Linear Formula (CH3CH2CH2CH2)2NH

  • Molecular Weight 129.24

  •  Beilstein/REAXYS Number 506001

  •  EC Number 203-921-8

  •  MDL number MFCD00009429

Purchase

Properties

vapor density   4.46 (vs air)
vapor pressure   1.9 mmHg ( 20 °C)
assay   ≥99%
autoignition temp.   594 °F
expl. lim.   10 %
refractive index   n20/D 1.417 (lit.)
bp   159 °C (lit.)
mp   −62 °C (lit.)
density   0.767 g/mL at 25 °C (lit.)
SMILES string   CCCCNCCCC
InChI   1S/C8H19N/c1-3-5-7-9-8-6-4-2/h9H,3-8H2,1-2H3
InChI key   JQVDAXLFBXTEQA-UHFFFAOYSA-N
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2248 3(8) / PGII
WGK Germany 
1
RTECS 
HR7780000
Flash Point(F) 
104 °F
Flash Point(C) 
40 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?