EMAIL THIS PAGE TO A FRIEND
240109 Sigma-Aldrich

Sorbic acid

≥99%

Synonym: α-trans-γ-trans-Sorbic acid, (E,E)-1,3-Pentadiene-1-carboxylic acid, 2,4-Hexadienoic acid, 2E,4E-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, C6:2n-2,4, NSC 35405, NSC 49103, NSC 50268

  • CAS Number 110-44-1

  • Linear Formula CH3CH=CHCH=CHCOOH

  • Molecular Weight 112.13

  •  Beilstein Registry Number 1098547

  •  EC Number 203-768-7

  •  MDL number MFCD00002703

Purchase

Properties

vapor pressure   0.01 mmHg ( 20 °C)
InChI Key   WSWCOQWTEOXDQX-MQQKCMAXSA-N
assay   ≥99%
mp   132-135 °C (lit.)
antibiotic activity spectrum   fungi

Description

Biochem/physiol Actions

Sorbic acid, a conjugated polyunsaturated fatty acid, improves growth performance in swine, by enhancing insulin-like growth factor gene expression and secretion in hepatocytes .

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
WG2100000
Flash Point(F) 
260.6 °F
Flash Point(C) 
127 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S1626 Sorbic acid, ≥99.0%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?