• USA Home
  • 240443 - 1,2,4-Trichlorobenzene

EMAIL THIS PAGE TO A FRIEND
240443 Aldrich

1,2,4-Trichlorobenzene

≥99%

  • CAS Number 120-82-1

  • Empirical Formula (Hill Notation) C6H3Cl3

  • Molecular Weight 181.45

  •  Beilstein/REAXYS Number 956819

  •  MDL number MFCD00000547

Purchase

Properties

vapor density   >6 (vs air)
vapor pressure   1 mmHg ( 40 °C)
InChI Key   PBKONEOXTCPAFI-UHFFFAOYSA-N
assay   ≥99%
autoignition temp.   1060 °F
expl. lim.   6.6 %, 150 °F
refractive index   n20/D 1.571 (lit.)
bp   214 °C(lit.)
mp   16 °C (lit.)
density   1.454 g/mL at 25 °C (lit.)
Safety & Documentation
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?