EMAIL THIS PAGE TO A FRIEND
240524 Sigma-Aldrich

Adipic acid

≥99%

Synonym: ADI-PURE® LGA adipic acid, ADI-PURE® high purity adipic acid, Hexanedioic acid

  • CAS Number 124-04-9

  • Linear Formula HOOC(CH2)4COOH

  • Molecular Weight 146.14

  •  Beilstein/REAXYS Number 1209788

  •  EC Number 204-673-3

  •  MDL number MFCD00004420

  •  PubChem Substance ID 329752550

Purchase

Properties

vapor density   5 (vs air)
vapor pressure   1 mmHg ( 159.5 °C)
assay   ≥99%
autoignition temp.   788 °F
bp   265 °C/100 mmHg (lit.)
mp   151-154 °C (lit.)
SMILES string   OC(=O)CCCCC(O)=O
InChI   1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)
InChI key   WNLRTRBMVRJNCN-UHFFFAOYSA-N

Description

Legal Information

DuPont product

ADI-PURE is a registered trademark of Invista North America S.a.r.l.

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
WGK Germany 
1
RTECS 
AU8400000
Flash Point(F) 
384.8 °F
Flash Point(C) 
196 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

A26357 Adipic acid, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?