EMAIL THIS PAGE TO A FRIEND
241040 Sigma-Aldrich

Dimethyl malonate

≥99%

  • CAS Number 108-59-8

  • Linear Formula CH2(COOCH3)2

  • Molecular Weight 132.11

  •  Beilstein Registry Number 774261

  •  EC Number 203-597-8

  •  MDL number MFCD00008460

  •  PubChem Substance ID 24854457

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
vapor density   >1 (vs air)
InChI Key   BEPAFCGSDWSTEL-UHFFFAOYSA-N
assay   ≥99%
refractive index   n20/D 1.413(lit.)
bp   180-181 °C(lit.)
mp   −62 °C (lit.)
density   1.156 g/mL at 25 °C(lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
1
RTECS 
OO0950000
Flash Point(F) 
185 °F
Flash Point(C) 
85 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?