EMAIL THIS PAGE TO A FRIEND
241261 Sigma-Aldrich

Glycine

≥99%

Synonym: Aminoacetic acid, Aminoethanoic acid, Glycocoll

Purchase

Properties

Related Categories Allium cepa (Onion), Allium sativum (Garlic), Amino Acids & Derivatives, Cell Biology, Chemical Biology,
assay   ≥99%
pKa (25 °C)   (1) 2.35, (2) 9.60
  2.35
mp   240 °C (dec.) (lit.)
SMILES string   NCC(O)=O
InChI   1S/C2H5NO2/c3-1-2(4)5/h1,3H2,(H,4,5)
InChI key   DHMQDGOQFOQNFH-UHFFFAOYSA-N

Description

Biochem/physiol Actions

Inhibitory neurotransmitter in spinal cord, allosteric regulator of NMDA receptors.

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
MB7600000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number

Certificate of Origin (COO)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

G7126 Glycine, ReagentPlus®, ≥99% (HPLC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?