• USA Home
  • 242497 - Methyl trichloroacetate

EMAIL THIS PAGE TO A FRIEND
242497 Sigma-Aldrich

Methyl trichloroacetate

99%

  • CAS Number 598-99-2

  • Linear Formula CCl3CO2CH3

  • Molecular Weight 177.41

  •  Beilstein Registry Number 1756075

  •  EC Number 209-960-7

  •  MDL number MFCD00000794

  •  PubChem Substance ID 24854562

Purchase

Properties

Related Categories Building Blocks, C2 to C5, Carbonyl Compounds, Chemical Synthesis, Esters,
InChI Key   VHFUHRXYRYWELT-UHFFFAOYSA-N
assay   99%
refractive index   n20/D 1.455(lit.)
bp   152-153 °C(lit.)
density   1.488 g/mL at 25 °C(lit.)

Description

Packaging

25 g in glass bottle

Application

Methyl trichloroacetate was used for negative staining of protein that allowed unmodified proteins to be recovered for biological studies or transblot for amino acid sequence.

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2533 6.1 / PGIII
WGK Germany 
3
Flash Point(F) 
161.6 °F
Flash Point(C) 
72 °C

Meet Synthia - Retrosynthesis Sofware
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?