• USA Home
  • 242683 - 1,2-Dichloro-3-iodobenzene

EMAIL THIS PAGE TO A FRIEND
242683 Sigma-Aldrich

1,2-Dichloro-3-iodobenzene

97%

Purchase

Properties

assay   97%
bp   123-126 °C/20 mmHg(lit.)
mp   35-36 °C (lit.)
Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Flash Point(F) 
235.4 °F
Flash Point(C) 
113 °C
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?