• USA Home
  • 243124 - 2-Isobutyl-3-methoxypyrazine

EMAIL THIS PAGE TO A FRIEND
243124 Sigma-Aldrich

2-Isobutyl-3-methoxypyrazine

technical grade, 70%

Purchase

Properties

grade   technical grade
InChI Key   UXFSPRAGHGMRSQ-UHFFFAOYSA-N
assay   70%
refractive index   n20/D 1.49(lit.)
density   0.99 g/mL at 25 °C(lit.)

Description

Other Notes

remainder 2-isobutyl-6-methoxypyrazine

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
WGK Germany 
3
Flash Point(F) 
176 °F
Flash Point(C) 
80 °C
Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

297666 2-Isobutyl-3-methoxypyrazine, 99%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?