• USA Home
  • 243183 - S,S′-(1,3-Propanediyl)bis(isothiouronium bromide)

EMAIL THIS PAGE TO A FRIEND
243183

S,S′-(1,3-Propanediyl)bis(isothiouronium bromide)

98%

  • CAS Number 5442-32-0

  • Empirical Formula (Hill Notation) C5H13BrN4S2

  • Molecular Weight 273.22

  •  MDL number MFCD00009697

Purchase

Properties

assay   98%
SMILES string   Br.Br.NC(=N)SCCCSC(N)=N
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?