EMAIL THIS PAGE TO A FRIEND
245151 Aldrich

2-Pentanone

90%

Synonym: Methyl propyl ketone

  • CAS Number 107-87-9

  • Linear Formula CH3COCH2CH2CH3

  • Molecular Weight 86.13

  •  Beilstein Registry Number 506058

  •  MDL number MFCD00009400

  •  PubChem Substance ID 329752677

Purchase

Properties

vapor pressure   27 mmHg ( 20 °C)
InChI Key   XNLICIUVMPYHGG-UHFFFAOYSA-N
assay   90%
autoignition temp.   941 °F
expl. lim.   8.2 %
refractive index   n20/D 1.39(lit.)
bp   101-105 °C(lit.)
mp   −78 °C (lit.)
density   0.809 g/mL at 25 °C(lit.)

Description

Other Notes

remainder 4-methyl-2-pentanone

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1249 3 / PGII
WGK Germany 
1
Flash Point(F) 
53.6 °F
Flash Point(C) 
12 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?