• USA Home
  • 246964 - Magnesium chloride hexahydrate

EMAIL THIS PAGE TO A FRIEND
246964 Sigma-Aldrich

Magnesium chloride hexahydrate

99%, ACS reagent

Purchase

Properties

grade   ACS reagent
assay   99%
  99.0-102.0%
impurities   ≤0.005% insolubles
anion traces   nitrate (NO3-): ≤0.001%
  phosphate (PO43-): ≤5 ppm
  sulfate (SO42-): ≤0.002%
cation traces   Ba: ≤0.005%
  Ca: ≤0.01%
  Fe: ≤5 ppm
  K: ≤0.005%
  Mn: ≤5 ppm
  NH4+: ≤0.002%
  Na: ≤0.005%
  Sr: ≤0.005%
  heavy metals: ≤5 ppm
SMILES string   O.O.O.O.O.O.Cl[Mg]Cl
InChI   1S/2ClH.Mg.6H2O/h2*1H;;6*1H2/q;;+2;;;;;;/p-2
InChI key   DHRRIBDTHFBPNG-UHFFFAOYSA-L
Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
1
RTECS 
OM2975000

Documents

Certificate of Analysis (COA)

Please Enter a Lot Number
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?